Principaux documents
SIAL0165
Sigma-Aldrich
Sigma® cell culture dish
diam. × H 35 mm × 10 mm, tissue-culture treated, sterile, pack of 25 × 20 ea
About This Item
Code UNSPSC :
41121812
Nomenclature NACRES :
NB.22
Stérilité
sterile
Conditionnement
pack of 25 × 20 ea
Diam. × H
35 mm × 10 mm
Superficie
8 cm2
Type de liaison
Tissue Culture (TC)-treated surface
Description générale
  • Manufactured from optically-clear virgin polystyrene
  • Treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Lid supplied with vents to provide consistent gas exchange
Informations légales
Sigma is a registered trademark of Sigma-Aldrich Co. LLC
Les clients ont également consulté
Protocoles
Universal Transfection Reagent enables efficient nucleic acid delivery into various cells, compatible with different cell culture conditions.
Calcium phosphate transfection is a common method for the introduction of DNA into eukaryotic cells. This protocol can be optimized for use with a wide variety of cell types.