Principaux documents
GE28-9889-46
Sigma-Aldrich
XK 16/70 Column
1 EACH
861,00 €
861,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Changer de vue
1 EACH
861,00 €
About This Item
Code UNSPSC :
41115711
Nomenclature NACRES :
SB.52
861,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Technique(s)
LPLC: suitable
Description générale
XK empty columns are designed for liquid chromatography at low to medium pressure. The user-friendly design ensures trouble-free operation with fittings for direct connection to ÄKTA design instruments or other high-performance liquid chromatography systems. The product family is based on a robust design for high reproducibility and accurate results. XK columns are compatible with aqueous solutions and most organic solvents used in liquid chromatography of macromolecules.
Caractéristiques et avantages
Adapter QuickLock mechanism that facilitates column handling and cleaning
Cooling jacket for sensitive purifications
Fittings for direct connection to chromatographiy systems
Plunger design to ensure a homogeneous buffer distribution and reproducible packing
User friendly and robust empty columns, designed for liquid chromatography at low to medium pressure.
Protocoles
Column Packing and Preparation for Size Exclusion Chromatography