Principaux documents
Z670146
StainTray slide staining system
StainTray with black lid
Synonyme(s) :
humidity tray, immunohistochemistry staining tray, slide staining tray
1 EA
493,00 €
493,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
493,00 €
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
MA.01
493,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
black acrylonitrile-butadiene-styrene (ABS) lid
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Simport M920-2
Paramètres
-80-80 °C temp. range
L × H × l
38 cm × 4.5 cm × 24 cm
Description générale
  • Holds up to 20 slides
  • Drain plug for easy emptying
  • 4 rubber feet for greater stability
  • Stackable for save space
These robust hunidity chambers offer a more user friendly approach to immunohistochemistry staining. This tray is suitable for routine staining requiring a humidity chamber. Wells between rails holds water securely without splashing. The plastic rails are covered with a polymer strip for perfectly holding slides even if tray is held at an angle. Rails are raised not only to avoid water touching the slides but to make them more easily retrievable. The base will also excess stain solution dripping from the slides.