EP5305000614
Eppendorf® Concentrator Plus
basic device, with rotor F-45-48-11, AC/DC input 230 V, 50—60 Hz, EuroPlug
About This Item
Code UNSPSC :
40161701
Produits recommandés
Caractéristiques
refrigerated
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 5305000614
Catégories apparentées
Description générale
With its advanced heating technology, the Eppendorf Concentrator plus is a vacuum concentrator that facilitates fast, efficient, yet gentle concentration of DNA, RNA, nucleotides, proteins, and other liquid or wet samples. Its coated lid provides superior chemical resistance against many acids and organic solvents.The latest Concentrator plus is a vacuum centrifuge that provides an ergonomic, intuitive, and durable solution, fitting comfortably on any lab bench. The coated lid improves durability by providing chemical resistance against acids and organic solvents. The Eppendorf concentrator Concentrator plus is available in three configurations, designed to meet your individual laboratory needs:
- The basic Concentrator plus system for connection to an external vacuum pump
- The Concentrator plus complete system with an integrated, solvent-resistant, maintenance, and oil-free PTFE diaphragm vacuum pump and condensation trap
- The same Concentrator plus complete system, but with an extra vacuum port added so you can use the integrated pump separately without having to disconnect it
Application
centrifugation, concentration
Caractéristiques et avantages
- Extremely quiet operation <50 dB(A)
- Small footprint saves valuable bench space
- Wide variety of 15 different rotor options to accommodate tubes from 0.2 mL to 50 mL, microplates and PCR plates
- Motorless induction drive and chemical-resistant, stainless steel chamber for trouble-free operation
- Can be connected with most commercially available cold traps for use with vapors
- Coated lid with superior chemical resistance
Informations légales
Eppendorf is a registered trademark of Eppendorf AG