Documents
EP0030132882
Eppendorf® real-time PCR tube strips without caps
pack of 120 ea
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.11
Matériaux
conical bottom
white tube
Caractéristiques
graduations: no
Conditionnement
pack of 120 ea
Fabricant/nom de marque
Eppendorf® 0030132882
Technique(s)
PCR: suitable
Capacité
0.1 mL
Description générale
  • White wells for better reflection.
  • High mechanical stability.
  • Extremely thin walls for optimal heat transfer.
  • Certified free from human DNA, DNase, RNase and PCR inhibitors.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG