Principaux documents
EP0030132890
Eppendorf® Masterclear cap strips and real-time PCR tube strips
10 x 12 strips
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.11
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Stérilité
non-sterile
Fabricant/nom de marque
Eppendorf® 0030132890
Technique(s)
PCR: suitable
Volume de travail
0.1 mL
Description générale
Eppendorf® Masterclear Cap Strips and real-time PCR Tube Strips:
Real-time PCR Tube Strips:
  • White wells for better reflection.
  • High mechanical stability.
  • Extremely thin walls for optimal heat transfer.
Masterclear Cap Strips:
  • Inverted dome to avoid scratching of the optic surface.
  • Inverted dome to reduce volume of the tubes.
  • Optimized for maximum light transmission.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG