Principaux documents
EP0030124910
Eppendorf® FAST PCR-Tube Strips
caps flat, pack of 120 ea
120 EA
266,00 €
266,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
120 EA
266,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.11
266,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
cap (plug seal)
colorless cap
colorless tube
conical bottom
Stérilité
non-sterile
Caractéristiques
PCR clean
graduations: no
neck thread Push fit
caps flat
Conditionnement
pack of 120 ea
Fabricant/nom de marque
Eppendorf® 0030124910
Technique(s)
PCR: suitable
Capacité
0.1 mL
Description générale
The Eppendorf Fast PCR Tube Strips made of Polyethylene transfer heat faster than the tubes made of traditional plastics. The Fast PCR Tube Strips are particularly suited to scientists using Fast PCR Reagent Kits, scientists trying to speed up protocols or cyclers with high heating and cooling rates - such as the Eppendorf Mastercycler X50 family.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG