Principaux documents
CCRF-HSB-2
85112801, human blood, Lymphoblast
About This Item
Code UNSPSC :
41106514
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Nom du produit
CCRF-HSB-2, 85112801, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Source biologique
human blood
Conditionnement
tube of 5 μg 85112801-DNA-5UG
pkg of vial of cells 85112801-1VL
Mode de croissance
Suspension
Caryotype
2n = 46
Morphologie
Lymphoblast
Produits
Not specified
Récepteurs
Not specified
Technique(s)
cell culture | mammalian: suitable
Maladie(s) pertinente(s)
cancer
Conditions d'expédition
dry ice
Température de stockage
−196°C
Origine de la lignée cellulaire
Human Caucasian acute lymphoblastic leukaemia
Description de la lignée cellulaire
A T lymphoblastoid line obtained from the peripheral blood of an 11 year old Caucasian male with acute lymphoblastoid leukaemia which has been passaged eight times through new-born Syrian Hamsters. These may be grown to a high density in a spinner type suspension culture.
Profil d'ADN
STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 10,12
D16S539: 9,13
D5S818: 11,12
D7S820: 11,12
THO1: 9,10
TPOX: 8
vWA: 18,19
Milieu de culture
IDMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Procédure de repiquage
Maintain cultures between 5-8x100,000 cells/ml; 5% CO2; 37°C. Maximum cell density 2-3x1,000,000 cells/ml.
Autres remarques
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.