Principaux documents
1.30191
Millipore
HY-LiTE® 2/2A Battery Holder
Replacement battery holder for battery operation of HY-LiTE® 2/2A luminometers
1 UNIT
227,00 €
227,00 €
Date d'expédition estimée le10 mars 2025Détails
Demander des informations complémentaires
Changer de vue
1 UNIT
227,00 €
About This Item
Code UNSPSC :
41122409
Nomenclature NACRES :
NA.74
227,00 €
Date d'expédition estimée le10 mars 2025Détails
Demander des informations complémentaires
Gamme de produits
HY-LiTE®
Fabricant/nom de marque
HY-LiTE®
Application(s)
food and beverages
Compatibilité
for use with HY-LiTE® 2/2A instrument versions.
Description générale
Replacement or spare battery holder for HY-LiTE® 2/2A instruments. The battery holder is used to operate the luminometer on battery power using 4 sealed alkaline batteries (size Mignon, LR6, AA). (Batteries not included).
Application
For use as a spare part or replacement of the original battery holder included with the luminometer.
Informations légales
HY-LITE is a registered trademark of Merck KGaA, Darmstadt, Germany