Principaux documents
Z679925
Bio-Pure pipetting reservoir
capacity 100 mL
Synonyme(s) :
multichannel pipette reservoir, reagent reservoir
100 EA
121,00 €
121,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
100 EA
121,00 €
About This Item
Code UNSPSC :
41121522
Nomenclature NACRES :
NB.01
121,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
PET (Bio-degradable)
V-bottom bottom
white
Stérilité
sterile
Conditionnement
pack of 100 ea
Fabricant/nom de marque
Diversified Biotech REBP-1000
Capacité
100 mL
Description générale
Bio-Pure Reagent Reservoirs are made from recaputured PET that does not rely on additional fossil fuel-derived petrochemicals. In addition, reservoirs are sealed in bio-degradable bags and packed in recycled cardboard cases. As a result, Bio-Pure Reservoirs are a more eco-friendly alternative. "Trough within a trough" maximizes reagent recovery. A wide base and reinforced sidewalls avoids spills and provides stability. Gamma irradiated RNASE/DNASE free
Caractéristiques et avantages
  • Constructed from biodegradable PET material for environmentally conscious disposal.
  • Specialized "trough within a trough" design for optimal reagent recovery.
  • Pour-off spouts at all four corners and a wide base for improved stability and spill prevention.
  • Sterilized for safe and contamination-free usage.
Informations légales
Bio-Pure is a trademark of Diversified Biotech