Z618217
Büchi® rotary evaporator Model R-205
professional, plastic-coated glass, with cold-trap condenser assembly
About This Item
Code UNSPSC :
41100000
Produits recommandés
Matériaux
plastic-coated glass
Disponibilité
not available in USA and Canada
Description générale
Modular construction makes it possible to configure units from two evaporator models, R-200/205, two vacuum controller models, V-800/V-805, and three different glassware assemblies, all supplied with a standard 24/40 joint on the evaporator flask and a 35/20 ball joint on the receiving flask.
Model R-200 Evaporators:
Model R-205 Evaporators:
Evaporator with diagonal condenser assembly
For simple distillation of solvents and general use. Tap-water-cooled. Inlet feed tube for continuous feed of volumes exceeding flask capacity.
Evaporator with cold-trap condenser assembly
Two-piece cold-trap condenser style is ideal for low-boiling solvents. Uses mechanical refrigeration or dry ice and acetone.
Evaporator with vertical condenser assembly
For distilling high-boiling solvents (>100°C). Tap-water-cooled. Inlet feed tube for continuous feed of volumes exceeding flask capacity.
3-STEP SELECTION GUIDE
1. Choose Glassware
Diagonal condenser, Vertical condenser, Cold-trap condenser
2. Choose Evaporator Model
R-200 Fully-equipped base model
R-205 Base model, plus digital vapor temperature display
3. Choose Level
BASIC Features of the selected evaporator model.
ADVANCED Features of the selected evaporator model, plus integrated, semi-automatic V-800 vacuum controller. Remote rotation speed and servo-jack control optional.
PROFESSIONAL Features of the selected evaporator model, plus integrated, programmable V-805 vacuum controller. Remote rotation speed and servo-jack control optional.
Model R-200 Evaporators:
- 33% smaller footprint than previous models
- Chemically-resistant PTFE/glass vacuum-sealing system lasts up to five years
- Space-saving modular design lets you add integrated vacuum controllers and run preset time/pressure gradients
- Motorized, quick-action servo-jack raises and lowers glassware effortlessly
- Wide-range, PTFE-coated 4L heating bath, model B-490, with digital temperature display, uses oil or water, 20 to 180°C (68 to 356°F)
- Enhanced safety: insulated heating bath, Plastic+Glass-coated glassware, and optional remote control and safety shield
- Process samples from 50 mL to 3 L
Model R-205 Evaporators:
- Same as R-200, plus digital vapor temperature display
Evaporator with diagonal condenser assembly
For simple distillation of solvents and general use. Tap-water-cooled. Inlet feed tube for continuous feed of volumes exceeding flask capacity.
Evaporator with cold-trap condenser assembly
Two-piece cold-trap condenser style is ideal for low-boiling solvents. Uses mechanical refrigeration or dry ice and acetone.
Evaporator with vertical condenser assembly
For distilling high-boiling solvents (>100°C). Tap-water-cooled. Inlet feed tube for continuous feed of volumes exceeding flask capacity.
3-STEP SELECTION GUIDE
1. Choose Glassware
Diagonal condenser, Vertical condenser, Cold-trap condenser
2. Choose Evaporator Model
R-200 Fully-equipped base model
R-205 Base model, plus digital vapor temperature display
3. Choose Level
BASIC Features of the selected evaporator model.
ADVANCED Features of the selected evaporator model, plus integrated, semi-automatic V-800 vacuum controller. Remote rotation speed and servo-jack control optional.
PROFESSIONAL Features of the selected evaporator model, plus integrated, programmable V-805 vacuum controller. Remote rotation speed and servo-jack control optional.
Informations légales
Büchi is a registered trademark of Büchi Labortechnik AG
Remplacé(e)(s) par