Principaux documents
Z508233
Sigma-Aldrich
Quick-release clamps
without extension, for flange 100 mm
Synonyme(s) :
clamps, quick release clamp
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.43
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Fabricant/nom de marque
Ace Glass 651725
Description générale
SS clamps for use with reaction flasks and heads with a 100 mm Schott flange. The extension rod can be clamped to the support frame and helps stabilize the reaction assembly.