Principaux documents
Z261327
Stainless steel 316 syringe needle, noncoring point
gauge 18, L 12 in.
Synonyme(s) :
Non-coring needle
1 EA
203,00 €
203,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
203,00 €
About This Item
Code UNSPSC :
42142600
Nomenclature NACRES :
NB.03
203,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Description
Micro-Mate Hub with 316 Stainless Steel Tubing
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Popper 9861
Longueur
12 in.
Calibre
18
Description générale
Specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. Female Luer hubs and needles are made of 316 SS. Noncontaminating, easy to clean and resistant to corrosion.
Micro-Mate Hub with 316 Stainless Steel Tubing