Documents
Z125687
Sigma-Aldrich
Ace three-neck round-bottom flasks
vertical, capacity 3,000 mL, Joints: ST/NS 24/40 (3)
Synonyme(s) :
bottom flask, flask round, round flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
round bottom flask
three-neck (Vertical)
Joint
Joints: ST/NS24/40 (3)
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper joint
Conditionnement
pack of 1
Fabricant/nom de marque
Ace Glass 6944-48
Capacité
3,000 mL