Principaux documents
TEM-CF400AU50
Sigma-Aldrich
Carbon Film Supported Gold Square Mesh
size 400 mesh, box of 50
Synonyme(s) :
CRBON FILM 400 MESH AU 50/BX
About This Item
Code UNSPSC :
41111761
Conditionnement
box of 50
Taille
400 mesh
Description générale
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Gold grid, 400 mesh.