Documents
SYNM240004
Synthware all-glass vacuum/inert gas manifold with solid high vacuum stopcocks
number of ports, 4, Hose Connections: Front-right, Rear-left
Synonyme(s) :
Synthware M240004, vacuum manifold, vacuum unit
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.51
Caractéristiques
number of ports 4
Fabricant/nom de marque
M240004
Informations légales
Synthware is a trademark of Kemtech America Inc.