Principaux documents
ESYNTH007
Sigma-Aldrich
SynLectro PTFE Electrode Holder
1 EACH
344,00 €
344,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Changer de vue
1 EACH
344,00 €
About This Item
Code UNSPSC :
81141502
Nomenclature NACRES :
NB.61
344,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Description générale
SynLectro PTFE Electrode Holder is part of the SynLectro Electrolysis Platform for synthetic electrochemistry. This electrode holder is designed to hold electrodes in firmly place to create a homogenous electric field. The PTFE holder prevents damage and scratching to the electrode surface - for use with soft electrode materials.
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Informations légales
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
Contenu apparenté
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.