Principaux documents
CLS9990100
Pyrex® ribbed watch glasses
O.D. ~100 mm
Synonyme(s) :
Corning® 9990
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
Corning 9990-100
D.E.
~100 mm
Description générale
Molded-in ribs allow vapors to escape during boiling to eliminate pressure buildup. Meets EPA requirements for methods 3005, 3010, 3020.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.