Principaux documents
BR91254
BRAND® glass beaker with spout, low form
volume 1000 mL, with graduation
Synonyme(s) :
Beakers, Glass
10 EA
212,00 €
212,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
10 EA
212,00 €
About This Item
Code UNSPSC :
41121803
Nomenclature NACRES :
NB.43
212,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
borosilicate glass 3.3
Caractéristiques
with graduation
Fabricant/nom de marque
BRAND 91254
Volume
1000 mL
Description générale
The BRAND® Glass Beaker with Spout is expertly crafted from Boro 3.3 glass, making it a reliable choice for laboratory applications. Designed in accordance with DIN EN ISO 3819 standards, this low-form beaker features clear graduation markings for precise measurement of liquids. The integrated spout allows for easy pouring, minimizing spills and enhancing accuracy during liquid transfer.
Autres remarques
Browse all glass beakers
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG