Principaux documents
BR759215
BRAND® UV cuvette micro
center H 8.5 mm, volume 70-850 μL, pack of 100 ea (individually wrapped)
Synonyme(s) :
Cuvettes, UV-Cuvettes
100 EA
61,80 €
61,80 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
100 EA
61,80 €
About This Item
Code UNSPSC :
41121813
Nomenclature NACRES :
SB.32
61,80 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pack of 100 ea (individually wrapped)
Fabricant/nom de marque
BRAND 759215
H centrale
8.5 mm
Volume
70-850 μL
Catégories apparentées
Description générale
  • BRAND® UV cuvette micro is manufactured under controlled room conditions.
  • Ideal for the determinations in water analysis, chemistry, and life science applications.
  • Cuvette material offers high chemical resistance to most polar solvents, acids, and alkaline solutions.
  • Clear, clean optical path with an indication of optical path orientation.
  • Eliminates the risk of cross-contamination.
  • Grouped by mold cavity number.
  • An economical alternative to Quartz cuvettes.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG