Principaux documents
BR36945
BRAND® volumetric flask BLAUBRAND® USP 27 conformity certified
capacity 20 mL, neck joint: ST/NS 10/19, polypropylene stopper
Synonyme(s) :
BLAUBRAND®, USP, Volumetric flasks
2 EA
39,90 €
39,90 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
2 EA
39,90 €
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
39,90 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
borosilicate glass
polypropylene stopper
Joint
joint: ST/NS neck10/19
Caractéristiques
accuracy class A
Conditionnement
pack of 2
Fabricant/nom de marque
BRAND 36945
Capacité
20 mL
Description générale
The BRAND® volumetric flask BLAUBRAND® USP 27 conformity certified is constructed from high-quality Boro 3.3 glass and adheres to DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring precise liquid measurement for various laboratory applications. Each flask is accompanied by a USP batch certificate, confirming its compliance with stringent quality standards. This volumetric flask is an essential tool for accurate analytical work, making it suitable for laboratories that require reliable and certified liquid handling solutions.
Caractéristiques et avantages
  • BLAUBRAND® 20 mL volumetric flasks offer precise measurement of liquids with high accuracy even for partial volumes.
  • Computer-controlled systems guarantee the possible precision for fully automated production processes.
  • Individually calibrated for the best technical perfection.
  • USP volumetric instruments comply with the Class A error limits required by the United States Pharmacopeia.
  • The flask can be heated up to 250°C in a drying cabinet or sterilizer without any permanent volume changes.
  • The use of specially developed high-quality printing inks ensures a long service life.
  • The blue enamel used for BLAUBRAND® volumetric instruments offers an excellent color contrast. It represents the optimum combination of resistance and readability.
Informations légales
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG