BAH369460000
SP Bel-Art® Sterileware™ Long Handled Sampling Spoon
white polystyrene, nominal capacity 14.79 mL , 3tsp, L × W 17.8 cm (7 in.) × 1.5625 in. (40 mm) mL, sterile, pack of 200 ea (individually wrapped)
Synonyme(s) :
Lab Scoop, dispensing tool, spoon
253,00 €
About This Item
253,00 €
Produits recommandés
Matériaux
white polystyrene
Stérilité
sterile
Conditionnement
pack of 200 ea (individually wrapped)
Fabricant/nom de marque
Bel-Art H36946-0000
L × l
17.8 cm (7 in.) × 1.5625 in. (40 mm) mL
Capacité nominale
14.79 mL , 3tsp
Catégories apparentées
Description générale
Caractéristiques et avantages
- Long handle reaches into jars, bottles and other confined areas
- Minimizes the potential of personal contact with sampled material
- Sterile, FDA grade polystyrene safe for food, drug and cosmetic use
- Individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing
Note: Sterilized products are not returnable.