Principaux documents
BAH195590000
SP Bel-Art® Straight Tubing Connectors
tubing I.D. 0.5-0.5 in. (4.7-12.7 mm)
Synonyme(s) :
connectors, tubing, tubing coupler
12 EACH
68,10 €
68,10 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
12 EACH
68,10 €
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
68,10 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pack of 12 ea
Fabricant/nom de marque
Bel-Art H19559-0000
L × l
3.375 in. (85.7  mm) × 0.8125 in. (20.6 mm)
D.I. du tuyau
0.5-0.5 in. (4.7-12.7 mm)
Description générale
These straight fittings ensure a tight, secure connection between lengths of tubing having the same inner diameter (I.D.).
Caractéristiques et avantages
  • Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases.
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Questions
  1. does this item contain several different adaptors with different diameters or just on etype
    1 answer
    1. This product includes 12 tubing connectors of the same size as shown below.