Principaux documents
BAH195700000
SP Bel-Art® Tubing Connector Set
22 stepped and straight tubing connectors
Synonyme(s) :
connectors, tubing, tubing coupler
22 EACH
92,20 €
92,20 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
22 EACH
92,20 €
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
92,20 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pack of 22 ea
Fabricant/nom de marque
Bel-Art H19570-0000
Description générale
This assortment of 22 stepped and straight tubing connectors comes in handy around the laboratory, and contains fittings compatible with tubing from 3.2 to 12.7mm (⅛ to ½").
Caractéristiques et avantages
  • Chemical-resistant polypropylene for use with a wide range of liquids or gases
  • Includes 12 Stepped Tubing Connectors and 10 Straight Tubing Connectors
  • FDA-grade polypropylene for direct contact with food, drugs, and cosmetics
  • Autoclavable
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP