Skip to Content
Merck
All Photos(1)

Key Documents

72812

Supelco

Ethyl 2-cyano-3,3-diphenylacrylate

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€60.30

€60.30


Please contact Customer Service for Availability


Select a Size

Change View
100 MG
€60.30

About This Item

Linear Formula:
(C6H5)2C=C(CN)CO2C2H5
CAS Number:
Molecular Weight:
277.32
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.24

€60.30


Please contact Customer Service for Availability

grade

analytical standard

Quality Level

Assay

≥99% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

174 °C/0.2 mmHg (lit.)

mp

≥95-100 °C
97-99 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CCOC(=O)\C(C#N)=C(/c1ccccc1)c2ccccc2

InChI

1S/C18H15NO2/c1-2-21-18(20)16(13-19)17(14-9-5-3-6-10-14)15-11-7-4-8-12-15/h3-12H,2H2,1H3

InChI key

IAJNXBNRYMEYAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service