Skip to Content
Merck
All Photos(1)

Key Documents

MMRECVERY

Millipore

MILLIFLEX OASIS® filtration unit

volume 100 mL

Synonym(s):

Filtration apparatus, Filtration device, Filtration funnel, Filtration system

Sign Into View Organizational & Contract Pricing

Select a Size

24 EACH
€124.00

€124.00


Estimated to ship on19 May 2025Details


Request more information

Select a Size

Change View
24 EACH
€124.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

€124.00


Estimated to ship on19 May 2025Details


Request more information

material

styrene-butadiene device

Quality Level

Agency

EP, USP, JP
EP 2.6.12
JP 4.05 part 1
USP 61

sterility

electron beam irradiated

feature

ready-to-use

packaging

pack of 24

manufacturer/tradename

Milliflex Oasis®

technique(s)

: suitable using growth promotion accessory

volume

100 mL

suitability

suitable for (suitable for solid cultivation)

application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® System

General description

The MILLIFLEX OASIS® filtration unit without membrane is designed specifically for the growth promotion test of the MILLIFLEX OASIS® culture media. This unit allows you to test the media performance under the same routine conditions.

Application

The Milliflex Oasis® filtration unit without a membrane is utilized for efficient bioburden testing and is designed for the growth promotion test.

Features and Benefits

  • Simple funnel transfer to the pump
  • Sample protection with the hinged lid on the funnel
  • Accurate sample volume with the clear funnel, visible level indicators, and hydrophobic material
  • Optimal, touch-free membrane to culture media transfer
  • Effortless funnel separation

Packaging

  • Units are packed by8 on 2 detachable trays.
  • Each box contains 24 units: 3 bags of 8 units.

Legal Information

MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service