Skip to Content
Merck
All Photos(1)

Key Documents

GERPN3688

Amersham AlkPhos Direct Hybridization Buffer

Cytiva RPN3688, pack of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.83

packaging

pack of 1 ea

manufacturer/tradename

Cytiva RPN3688

Related Categories

General description

AlkPhos Direct Labeling and Detection Systems are based on the rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase. CDP-Star or ECF detection can be used. Denatured or single-stranded DNA or RNA probe is mixed with the labelling buffer and alkaline phosphatase. The probe is hybridized to the blot using the specially formulated AlkPhos Direct Hybridization buffer, which stabilizes the activity of the enzyme. After a 15-minute pre-hybridization step, hybridization with probe is performed overnight at the required temperature. For higher target amounts, a 2–4 hour hybridization may be sufficient.

Features and Benefits

  • Fast and easy to use system suitable for the majority of routine blotting applications, using either DNA, RNA or oligonucleotide probes.
  • Alk Phos Direct system has less components than other non-radioactive systems, making it simpler to use as less components need to be removed during the stripping procedure.
  • Probe removal is more effective with less damage to the membrane.
  • Optimized protocol has been developed capable of labelling probes down to 17 base pairs in length.
  • 3–4 hours of time savings compared with conventional, indirect methods are achievable as a result of the rapid and simple labelling reaction.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

AlkPhos Direct
This product is protected by patent numbers US 5,939,257 and EP 770678 owned by Cytiva Bio-Sciences Corp. and equivalent patents and patent applications in other countries.

Hazard Statements

Precautionary Statements

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service