Key Documents
22406
Supelco
Puller/Inserter Tool
pkg of 2 ea
Select a Size
2 EA
€40.50
€40.50
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
2 EA
€40.50
About This Item
UNSPSC Code:
12352200
€40.50
Please contact Customer Service for Availability
Request a Bulk Order
packaging
pkg of 2 ea
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
Simplifies the task of inserting or removing wool or foam plugs when working with GC inlet liners, packed GC columns, solvent desorption tubes, thermal desorption tubes, and purge traps. The forked end is used to feed the plug into tubing, leaving it cleanly when the tool is removed. The hooked end will not disrupt the packing material when the plug is removed. Made of stainless steel, this tool can be used with 1 - 4 mm I.D. tubing (glass, metal, and PTFE).
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 2
1 of 2