Skip to Content
Merck
All Photos(1)

Key Documents

22713

Swagelok® Miniature Quick Connect Body Assembly

Swagelok® QM2-B-200, 1/8 in. Swagelok fitting, stainless steel

Synonym(s):

Swagelok® QM2-B-200,SS MINI QK CONN 1/8" EA

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€158.00

€158.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€158.00

About This Item

UNSPSC Code:
41115717
eCl@ss:
32035302

€158.00


Please contact Customer Service for Availability

Request a Bulk Order

material

stainless steel

packaging

pkg of 1 ea

manufacturer/tradename

Swagelok® QM2-B-200

fitting

1/8 in. Swagelok fitting

compatibility

for use with miniature Quick Connects stem assembly (22711-U)

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emre Balta et al.
Nature communications, 10(1), 4073-4073 (2019-09-11)
Several antitumor therapies work by increasing reactive oxygen species (ROS) within the tumor micromilieu. Here, we reveal that L-plastin (LPL), an established tumor marker, is reversibly regulated by ROS-induced thiol oxidation on Cys101, which forms a disulfide bridge with Cys42.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service