22139-U
On/Off Throttling Valve
stainless steel, 1/8 in.
Synonym(s):
Swagelok® 2P4T,SS THROTTLING VALVE 1/8" EA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
stainless steel
fitting
1/8 in.
Looking for similar products? Visit Product Comparison Guide
General description
Throttling valve′s on and off settings determines whether or not fluid flows past the valve. It is much more efficient than the toggle on/off valve, as it provides better flow control between 0 and 100% flow and has a larger baseline pressure drop. There are many styles of throttling valve like globe valve and butterfly valve.
Nuts and ferrules included.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Process Plant Equipment: Operation, Control, and Reliability.
Technology & Engineering, 401-402 (2012)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service