Skip to Content
Merck
All Photos(1)

Key Documents

21964

SGE Gas Tight Syringes, Luer Lock

1MDF-LL-GT, volume 1000 μL, needle size (not included)

Synonym(s):

SYRINGE SGE 1ML GAS TIGHT LUER LOCK, EA

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€85.80

€85.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€85.80

About This Item

UNSPSC Code:
41122001
eCl@ss:
32070500

€85.80


Please contact Customer Service for Availability

Request a Bulk Order

material

glass syringe

needle size

(not included)

feature

needle type Luer lock

packaging

pkg of 1 ea

manufacturer/tradename

SGE 008025

volume

1000 μL

Looking for similar products? Visit Product Comparison Guide

Application

SGE Gas Tight Syringes, Luer Lock was used:
  • for collection of estuary water sediment, in an experimental study done to assay the anammox reaction occurring in estuary sediments.
  • to sub-sample, 15N-N2 and N2O sample gas in head-space, during measurement of annamox and denitrification in intact sediment cores.[1]
  • with combination of PCC, which is basically a cell homogenizer, during the isolation of mitochondria, in an experimental investigation done to study the progression of human diseases.[2]
  • for the introduction of the sample in to the GASIS (gas sampling and injection system) inlet, during the compound-specific δD and δ13C gaseous mixture analysis by mass spectrometry.[3]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A semi-automated method for isolating functionally intact mitochondria from cultured cells and tissue biopsies
Scmitt S
Analytical Biochemistry, 443 (1), 66-74 (2013)
Direct measurement of anaerobic ammonium oxidation (anammox) and denitrification in intact sediment cores.
Trimmer M
Marine Ecology Progress Series, 326, 37-47 (2006)
Versatile inlet system for on-line compound-specific dD and d13C gas chromatography-oxidation/reduction-isotope ratio mass spectrometry analysis of gaseous mixtures.
Henning M
Rapid Communications in Mass Spectrometry, 21 (!4), 2269-2272 (2007)

Questions

  1. Do you have a picture of this syringe? What is the internal diameter?

    1 answer
    1. The picture of the syringe is attached. The I.D. of the syringe is not disclosed. But there are also alternative products like 29638-U and 509388 with needle I.D. values of 0.37mm and 0.32 mm, respectively.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service