Skip to Content
Merck
All Photos(1)

Key Documents

21533

SGE Gas Tight Syringes, Luer Lock

100MR-LL-GT, volume 100 mL, needle size (not included)

Synonym(s):

SYRINGE SGE 100MR-LL-GT, 100ML

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€433.00

€433.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€433.00

About This Item

UNSPSC Code:
41122001

€433.00


Please contact Customer Service for Availability

Request a Bulk Order

material

glass syringe

needle size

(not included)

feature

needle type Luer lock

packaging

pkg of 1 ea

manufacturer/tradename

SGE 009760

volume

100 mL

Looking for similar products? Visit Product Comparison Guide

Application

SGE gastight Luer lock syringe was used to collect gas samples from the slightly pressurized headspace, while measuring anaerobic ammonium oxidation.[1] It was also used to collect gas samples of the headspace gas, during an experimental procedure related to increase in the occurrence of the anammox reaction in estuarine sediments.[2] It may also be used for introduction of samples to the gas sampling and injection system (GASIS) inlet, in a study involving versatile inlet system for on-line compound-specific δD and δ13C gas chromatography-oxidation/reduction-isotope ratio mass spectrometry analysis of gaseous mixtures.[3]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark Trimmer et al.
Applied and environmental microbiology, 69(11), 6447-6454 (2003-11-07)
Until recently, denitrification was thought to be the only significant pathway for N(2) formation and, in turn, the removal of nitrogen in aquatic sediments. The discovery of anaerobic ammonium oxidation in the laboratory suggested that alternative metabolisms might be present
Widespread occurrence of the anammox reaction in estuarine sediments.
Nicholls JC and Trimmer M.
Aquatic Toxicology (Amsterdam, Netherlands), 55 (2), 105-105 (2009)
Mark Henning et al.
Rapid communications in mass spectrometry : RCM, 21(14), 2269-2272 (2007-06-20)
Compound-specific deltaD and delta13C analyses of gas mixtures are useful indicators of geochemical and environmental factors. However, the relative concentrations of individual components in gas mixtures (e.g., H2, CO2, methane, ethane, propane, i-butane, n-butane) may vary over several orders of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service