Skip to Content
Merck
All Photos(1)

Key Documents

R4777

Sigma-Aldrich

Anti-Ran antibody,Mouse monoclonal

clone ARAN1, purified from hybridoma cell culture

Synonym(s):

Anti-ARA24, Anti-Gsp1, Anti-TC4

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€1,030.00

€1,030.00


Please contact Customer Service for Availability


Select a Size

Change View
200 μL
€1,030.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€1,030.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

ARAN1, monoclonal

form

buffered aqueous solution

species reactivity

human, Xenopus, bovine, mouse, hamster

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.5-1 μg/mL using total extract of Jurkat cells

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAN(5901)
mouse ... Ran(19384)

Related Categories

General description

Monoclonal Anti-Ran (mouse IgG2b isotype) is derived from the ARAN1 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BDF1 mice immunized with denatured recombinant human Ran. Ran is a small guanine trinucleotide phosphatase (GTPase) of the Ras superfamily that like other GTPases exist in two states: bound to guanine trinucleotide phosphate (GTP) or to guanine dinucleotide phosphate (GDP). The bound form is localized to the nucleus.

Immunogen

denatured recombinant human Ran.

Application

Monoclonal Anti-Ran antibody produced in mouse has been used in:
  • western blotting
  • immunoprecipitation
  • enzyme linked immunosorbent assay (ELISA)
  • immunocytochemistry

Biochem/physiol Actions

Ran plays an important function in the directionality of nuclear transport. In this form it is important for the termination of the import reaction which is mediated by a complex of the cargo protein with importin-β and its adapter protein importin-α. The binding of RanGTP to importin-β causes the release of the cargo protein from the importin-α/importin-β complex. The import receptor (importin-β or transportin) is then exported out of the nucleus as a RanGTP complex and is released into the cytoplasm when RanGTP is converted to RanGDP by Ran-GAP1 (GTPase-activating protein). Export receptors respond in the opposite way by binding cargo with much higher affinity in the presence of RanGTP and exporting them out of the nucleus as RanGTP complexes. The exported cargo is released when Ran GAP1 converts RanGTP to RanGDP. In order to maintain the import/export cycle, Ran has to return to the nucleus. This is achieved by a small protein called nuclear transport factor 2 (NTF2) also known as p10, that binds RanGDP in the cytoplasm, and delivers it back into the nucleus. In the nucleus RanGDP encounters RanGEF (guanine nucleotide exchange factor) and undergoes nucleotide exchange to become RanGTP.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein import into nuclei: association and dissociation reactions involving transport substrate, transport factors, and nucleoporins
Rexach M and Blobel G
Cell, 83(5), 683-692 (1995)
Insights into the molecular mechanism of nuclear trafficking using nuclear transport factor 2 (NTF2)
Stewart M
Cell Structure and Function, 25(4), 217-225 (2000)
SCF beta-TRCP controls oncogenic transformation and neural differentiation through REST degradation
Westbrook TF, et al.
Nature, 452(7185), 370-370 (2008)
Nuclear protein import
Gorlich D
Current Opinion in Cell Biology, 9(3), 412-419 (1997)
Ran, a GTPase involved in nuclear processes: its regulators and effectors
Avis JM and Clarke PR
Journal of Cell Science, 109(10), 2423-2427 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service