Skip to Content
Merck
All Photos(1)

Key Documents

P8240

Sigma-Aldrich

REDExtract-N-Amp PCR ReadyMix for Blood

12 mL sufficient for 1000 amplifications

Sign Into View Organizational & Contract Pricing

Select a Size

12 ML
€1,480.00

€1,480.00


Please contact Customer Service for Availability


Select a Size

Change View
12 ML
€1,480.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.55

€1,480.00


Please contact Customer Service for Availability

Quality Level

usage

sufficient for 1000 reactions
12 mL sufficient for 1000 amplifications

feature

dNTPs included
hotstart

color

red

shipped in

wet ice

storage temp.

−20°C

General description

REDExtract-N-Amp PCR ReadyMix for Blood is a PCR master mix containing buffer, salts, dNTPs, and Taq polymerase. This 2x reaction mixture also contains JumpStart Taq antibody for specific hot start amplification and REDTaq® dye, an inert tracking dye to enable direct loading of PCR products onto an agarose gel for analysis. PCR ReadyMix is intended for use with Sigma′s Extract-N-Amp Blood PCR Kits. All Extract-N-Amp Kit include a volume of PCR ReadyMix sufficient for at least one PCR reaction per extraction. However, where additional amplifications are required, supplemental PCR ReadyMix may be needed.

Application

REDExtract-N-Amp PCR ReadyMix for Blood has been used for polymerase chain reaction (PCR).[1]

Features and Benefits

  • Additional loading dye is not required as the PCR reaction mix contains REDTaq® dye for direct and convenient loading onto an agarose gel for analysis
  • Hotstart feature allows for room temperature set-up without premature Taq DNA polymerase activity
  • Ready-mix enables time-saving procedures
  • Reduced risk of contamination from multiple pipet steps and increased reproducibility

Legal Information

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
JumpStart is a trademark of Sigma-Aldrich Co. LLC
REDExtract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
REDTaq is a registered trademark of Merck KGaA, Darmstadt, Germany
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sofia Lejon Crottet et al.
Transfusion and apheresis science : official journal of the World Apheresis Association : official journal of the European Society for Haemapheresis, 50(2), 169-174 (2014-04-01)
Starting in 2013, blood donors must be tested at least using: (1) one monoclonal anti-D and one anti-CDE (alternatively full RhCcEe phenotyping), and (2) all RhD negative donors must be tested for RHD exons 5 and 10 plus one further

Protocols

This procedure is for the determination of Peroxidase enzymatic activity using Pyrogallol as the substrate.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service