Aluminimum self adhesive foil seal has a strong acrylic adhesive which produces a seal of high integrity. Seal is pierceable and peelable and suitable for high temperature application.
Application
PCR
Incubation
Storage
Features and Benefits
Pierceable, when pierced, the foil tears in an irregular manner which prevents the formation of a vacuum
Peelable
Perforated end tabs for easy application and removal by peeling
Seal integrity range: -80°C to 120°C
Suitable for cutting to fit part plates
Legal Information
LabRoll is a trademark of IST Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.