Skip to Content
Merck
All Photos(1)

Key Documents

EP0030122313

Eppendorf® Tubes 5.0 mL

PCR clean, 5.0 mL, colorless, pkg of 200 tubes (2 bags of 100 tubes), with screw cap

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing

Select a Size

200 EA
€68.90

€68.90


Please contact Customer Service for Availability


Select a Size

Change View
200 EA
€68.90

About This Item

UNSPSC Code:
41121703
NACRES:
NB.22

€68.90


Please contact Customer Service for Availability

material

conical bottom polypropylene

sterility

non-sterile

feature

PCR clean
with screw cap

packaging

pkg of 200 tubes (2 bags of 100 tubes)

manufacturer/tradename

Eppendorf® 0030122313

measuring range

5.0 mL

parameter

25000 × g max. RCF

D

17 mm

color

colorless

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf 5.0 mL tube′s conical shape is designed to be compatible with most accessories for common conical 15 mL tubes. This allows you to continue using many of your existing adapters and racks!Eppendorf′s "missing link": the Eppendorf Tube 5.0 mL, fills the gap between existing tube versions and enables the simple and safe processing of sample volumes up to 5.0 mL.

Application

Eppendorf® Tubes 5.0 mL is suitable for:
  • Easy and safe cell and tissue lysis
  • Preparing and storing buffers and master mixes
  • Higher yields of DNA samples from less quantity of source material
  • Safe and convenient cell culture applications

Features and Benefits

  • The screw cap is flattened and has grooved sides that enable user-friendly handling and optimal grip
  • The screw cap can withstand a broad operating temperature range from -86 °C – 100 °C
  • Ensures reliable test results as it is made up of high-quality transparent polypropylene that is free of plasticizers, biocides, or mold release agents
  • Highly safe and stable for centrifugation up to 25,000 x g that ensures fast, reliable, and efficient protocols
  • The conical bottom shape ensures compatibility with accessories for 15mL conical tubes
  • For practical and cost-effective lab procedures
  • Less storage space required
  • Easier processing of samples up to 5.0mL

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service