Skip to Content
Merck
All Photos(1)

Key Documents

EP0030120086

Eppendorf® Safe-Lock micro test tubes

capacity 1.5 mL, colorless, pack of 1000 ea

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€52.50

€52.50


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€52.50

About This Item

UNSPSC Code:
41121702
NACRES:
NB.11

€52.50


Please contact Customer Service for Availability

material

cap (snap cap)
colorless
colorless cap
colorless tube
conical bottom
polypropylene cap
polypropylene tube

feature

graduations
neck thread Push fit

packaging

pack of 1000 ea

manufacturer/tradename

Eppendorf® 0030120086

capacity

1.5 mL

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf® Safe-Lock micro test tubes protect your sample with excellent centrifugation stability, feature optimized sealing properties, and reliably prevent sample evaporation.

Features and Benefits

  • Eppendorf® Safe-Lock lid prevents unintentional lid opening during incubation and storage ensuring the highest sample protection. This feature is especially important when working with precious samples, or buffers containing detergents such as SDS.
  • Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes.
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
  • g-Safe®: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples.
  • Precise lid sealing for minimal evaporation rates during long-term storage.
  • Large frosted lid and surface on the side for easy labeling.
  • Ensured functionality from -86 °C to 100 °C.
  • Autoclavable when open (121 °C, 20 min).
  • The hinged Eppendorf® Safe-Lock lid provides additional protection against unintentional lid opening during incubation and storage.
  • g-Safe®:Provides great centrifugation stability and prevents sample loss due to tube breakage
  • The optimized sealing properties ensures safety when working with hazardous samples and minimal evaporation rates during long-term storage.
  • The large frosted lid and wide side-surface area make labeling convenient.
  • Functions between temperatures -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service