Skip to Content
Merck
All Photos(1)

Key Documents

AXYTF1000RS

Pipette filter tips

universal fit, racked, size 1000 μL, pkg of 5x1000tips/cs, sterile

Synonym(s):

Filtered Pipette Tips, Pipette Filters

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€578.00

€578.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€578.00

About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume:
1000 μL
sterility:
sterile
feature:
DNA free
RNase free
barrier
endotoxin free
filter
packaging:
pkg of 5x1000tips/cs
manufacturer/tradename:
Axygen Inc TF-1000-R-S
Corning

€578.00


Please contact Customer Service for Availability

material

clear

sterility

sterile

feature

DNA free
RNase free
barrier
endotoxin free
filter

manufacturer/tradename

Axygen Inc TF-1000-R-S
Corning

packaging

pkg of 5x1000tips/cs

size

1000 μL

volume

1000 μL

Looking for similar products? Visit Product Comparison Guide

General description

Axygen® Filter Barrier tips offer a universal design compatible with various pipettes for ease of use. They are available in a rack format, allowing for convenient and organized storage. Additionally, unique filtration functions and reliable performance provide enhanced sample protection and contamination control during pipetting tasks.

Application

Pipette filter tips have been used in:

  • microbiological and DNA amplification process[1][2]
  • diagnostics[3]

Features and Benefits

  • To hinder and block aerosols, the filter creates a barrier within the tip, preventing cross-contamination between samples
  • This polyethylene filter, devoid of DNA inhibitor, is robotically put into the tip to avoid nuclease contamination
  • To avoid contamination, filter tips are stored with lids in sealed-bottom racks
  • Sterile and free from RNase/DNase
  • Nonpyrogenic

Other Notes

Browse all pipette tips

Legal Information

Axygen is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Formation of Minimised Hairpin Template-transcribing Dumbbell Vectors for Small RNA Expression
Jiang X
Bio-protocol (2017)
Pathogenicity Assay of Penicillium expansum on Apple Fruits
Yong C, et al.
Bio-protocol (2107)
Kirsty J Dixon et al.
Bio-protocol, 7(9) (2017-07-21)
The subventricular zone (SVZ) in the mammalian forebrain contains stem/progenitor cells that migrate through the rostral migratory stream (RMS) to the olfactory bulb throughout adulthood. SVZ-derived explant cultures provide a convenient method to assess factors regulating the intermediary stage of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service