Skip to Content
Merck
All Photos(1)

Key Documents

NIST978A

Assay-isotopic standard for silver

NIST® SRM® 978a

Synonym(s):

Silver nitrate, Nitric acid silver(I) salt

Sign Into View Organizational & Contract Pricing

Select a Size

0.25 G
€658.00

€658.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
0.25 G
€658.00

About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€658.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

vapor density

5.8 (vs air)

isotopic purity

 107Ag
 108Ag

form

solid

packaging

pkg of 0.25 g

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

mp

212 °C (dec.) (lit.)

format

matrix material

SMILES string

[O-][N+]([O-])=O.[Ag+]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is certifed for use as an assay and isotopic standard. It has been prepared form high purity silver metal and high purity nitric acid. For more information, please refer to the SDS and COA.

SRM 978A_cert

SRM 978A_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Ag-107 and Ag-109

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service