Merck
Silver Compounds
Silver atomic symbol
Silver (Ag) is a bright white metal prized for its high electrical and thermal conductivity, as well as its reflectivity. It is commonly found in native silver form in the Earth’s crust and is often a byproduct of refining copper, gold, lead, and zinc. Silver compounds are versatile and find applications in electronics, particularly in the form of silver nanoparticles for their excellent conductivity. In pharmaceuticals, silver compounds, such as silver nitrate and silver sulfadiazine, exhibit potent antimicrobial properties used in wound care, implant coatings, and burn treatments. Silver nitrate is also utilized in photography and serves as a precursor in chemical synthesis.
Read more about
Products
Sort by Relevance
Showing 1-20 of 1005
Change View
Showing 1-20 of 1005
Compare
Product Number
Product Name
Product Description
Pricing
Silver as Catalyst
Silver catalysts are commonly used due to their high oxidation power and potential in the formation of silver complexes. Additionally, they function as silver activators, enhancing the electronegativity of other catalysts, such as gold. Both organic and inorganic synthesis benefit from the stoichiometric oxidation potential of silver compounds. Homogeneous silver-catalyzed organic transformations highlight the unique redox chemistry of silver, enabling the catalysis of reactions with high stereo- and regioselectivity. Silver catalysts efficiently mediate both intermolecular and intramolecular bond formations. Heterogeneous processes involving silver catalysis include NOx reduction and the catalytic oxidation of carbon monoxide (CO) to carbon dioxide (CO2). Silver(I) salts are also used in several silver-catalyzed nucleophilic addition reactions and organic transformations. Our portfolio offers a wide variety of high-quality silver catalysts for transition-metal catalysis in organic synthesis.
Silver Acetate
Silver acetate (AgC2H3O2) is a compound of silver ions (Ag+) coordinated with acetate ions (C2H3O2-) through ionic bonds. Commonly used in organic synthesis, it serves as a mild oxidizing agent. In analytical chemistry, its insolubility in water makes it a valuable reagent for detecting halides. Silver acetate also finds application in cigarette filters to reduce nicotine addiction by catalyzing the conversion of nicotine into an inactive form. Additionally, it has been explored for its potential antiviral properties, making it a subject of interest in medical research for combating viral infections. Moreover, it plays a role in a unique preparation method for highly reflective, conductive silvered polymer films, effectively catalyzing cycloaddition reactions of isocyanoacetates with various olefins.
Silver acetate is also a well-known precursor used in printed electronics. Particle-free "reactive inks", derived from complexes of silver acetate, have been reported to produce traces that approach the conductivity of bulk silver.
Silver Nitrate
Silver nitrate (AgNO3) is a versatile chemical compound with significant applications in both chemical and pharmaceutical industries. Highly soluble, it serves as a reagent in various laboratory reactions, including halide detection and the synthesis of other silver compounds. Functioning as a catalyst, silver nitrate facilitates the oxidation of aromatic, aliphatic, and conjugated aldehydes to their respective carboxylic acids, utilizing H2O2 as an oxidant. Furthermore, it aids in the air oxidation of benzylic and allylic alcohols, leading to the formation of corresponding aldehydes or ketones in the presence of Na2CO3. Additionally, silver nitrate plays a crucial role in the Friedel-Crafts acylation of benzene derivatives, contributing to the synthesis of corresponding ketones. It also supports the hydrolytic oxidation of organosilanes, resulting in the production of hydrogen.
In pharmaceuticals, silver nitrate is widely used for its potent antimicrobial properties. It is employed topically in wound care as a cauterizing agent to prevent infection and promote healing. Additionally, it is applied in treating eye conditions, like conjunctivitis in newborns, due to its antibacterial effects. In the production of medical devices, silver nitrate coatings prevent microbial colonization, enhance biocompatibility, and reduce infection risks. Silver nitrate serves as a precursor for synthesizing nanomaterials, silver composites, and silver compounds.
Silver Nanoparticles
Silver nanoparticles (Ag NPs) are vital in numerous industries due to their exceptional properties such as small size, large surface area, and quantum confinement effects. The catalytic, thermal, and optical properties of silver nanoparticles are significantly influenced by their size and shape. Silver nanoparticles exhibit antibacterial activity due to their surface structures and large surface-to-volume ratios compared to other nanomaterials. The interaction of Ag NPs with bacterial cells results in the accumulation of AgNPs in the cell wall, forming pits and causing cell death. Smaller Ag NPs are known to demonstrate more effective antibacterial activity than larger particles. Additionally, Ag NPs have been employed in biosensors due to the sharper and stronger plasmon resonance of silver, crucial for imaging systems. They serve as valuable tools in cancer diagnosis and treatment, functioning as drug carriers with the ability to target cancer cells, absorb light, and facilitate effective destruction through photothermal therapy. Interestingly, Ag NPs are utilized in the food industry due to their antibacterial properties.
Silver Nanowires
Silver nanowires (AgNWs) are 1-D silver-based nanostructures with diameters typically ranging from tens to hundreds of nanometers. They exhibit exceptional electrical, thermal, and optical properties, making them suitable materials for flexible conductive, optical, and anti-microbial applications such as touchscreen displays, solar cells, film heaters, medical imaging, and sterile clothing. Moreover, their large aspect ratio enables efficient light trapping in photovoltaic applications.
Beyond electronics, silver nanowires are utilized in sensors, catalysis, and biomedical applications, showcasing their versatility and potential for innovation across diverse industries. Their unique properties enable efficient encapsulation and transport of drugs, enhancing bioavailability and minimizing side effects. They also serve as platforms for biosensors, enabling rapid and sensitive detection of biomolecules for diagnostics and monitoring purposes.
Silver Oxide
Silver oxide (Ag2O) is a fine black or dark brown powder that is used in the synthesis of other silver compounds. In electronics, particularly in silver-oxide batteries, and silver-zinc batteries, it serves as the cathode material, offering high energy density and stability. In the pharmaceutical industry, silver oxide exhibits antimicrobial properties, making it a valuable ingredient in wound dressings and medical devices to prevent infections.
Silver oxide nanoparticles act as stable photocatalyst under visible and near-infrared light irradiation. Their photocatalytic activity in NIR light stems from a low band gap, less than 1.3 eV. Furthermore, the aggregation of Ag2O nanoparticles provides a large surface area and numerous crystal boundaries, enhancing the escape chance of photogenerated electrons and the contact likelihood of photogenerated holes with other materials. This results in outstanding photocatalytic activity and stability in produced Ag2O samples.
Silver Chloride
Silver chloride (AgCl) is a white crystalline compound widely used as a catalyst, antibacterial agent, photographic material, and ionic semiconductor material. The high photosensitivity of silver chloride makes it an important photocatalyst. It also acts as a catalyst in the preparation of 1,2,3-triazoles from alkynes and azides. Additionally, it serves as a precursor salt for synthesizing silver nanoparticles.
Electrochemically, silver chloride is employed in sensors and electrodes for its conductivity. In medicine, it applies in wound dressings and medical devices for antimicrobial properties, preventing infections.