Skip to Content
Merck
All Photos(3)

Key Documents

NIST987

Strontium carbonate

NIST® SRM® 987, isotopic standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€714.00

€714.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€714.00

About This Item

Linear Formula:
SrCO3
CAS Number:
Molecular Weight:
147.63
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€714.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 1 g

manufacturer/tradename

NIST®

density

3.7 g/mL at 25 °C (lit.)

format

neat

SMILES string

[Sr++].[O-]C([O-])=O

InChI

1S/CH2O3.Sr/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

LEDMRZGFZIAGGB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 987 consists of 1 g of strontium carbonate powder.

SRM 987 _SDS

SRM 987 _CoA

Application

Strontium carbonate standard reference material (SRM) is intended for use as a reference material for isotopic values, used to calibrate mass spectrometers.

Features and Benefits

  • Available with a certified values for the absolute strontium isotopic abundance ratios and the atom fractions of 88Sr, 87Sr, 86Sr and 84Sr.  
  • The NIST certificate is provided with maintenance, storage, and handling instructions.

Other Notes

Certified for the analytes listed below.
Strontium (88Sr, 87Sr, 86Sr, 84Sr)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service