Skip to Content
Merck
All Photos(1)

Key Documents

NIST3668

Mercury, perchlorate, and iodide in frozen human urine

NIST® SRM® 3668

Sign Into View Organizational & Contract Pricing

Select a Size

10 PKG
€1,770.00

€1,770.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 PKG
€1,770.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€1,770.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

liquid

packaging

pkg of Set (10) (5 ea conc)

manufacturer/tradename

NIST®

application(s)

clinical testing

General description

A unit of SRM 3668 consists of five vials of each of two levels.

Each vial contains 1.5 mL of urine.

SRM 3668_cert

SRM 3668_SDS

Application

NIST3668 Mercury, perchlorate, and iodide in frozen human urine standard reference material(SRM) is intended to use for validating analytical methods and measurements for the determination of toxic elements and anions in human urine.

Features and Benefits

  • Available with a certificate documenting NIST-certified mass concentration values of mercury, perchlorate, and Nitrate.
  • Reference values are expressed as mass concentration for iodine and thiocyanate.
  • The NIST certificate is provided with handling, storage,and use instructions.

Other Notes

Certified for the analytes listed below.
Mercury (Hg), Perchlorate (KClO4), Nitrate ( NO3-)

Reference/Informational value is provided for the following analytes.
Iodine (I), Thiocyanate (SCN ?), Creatinine (C4H7N3O)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service