Skip to Content
Merck
All Photos(1)

Key Documents

NIST2669

Arsenic species in frozen human urine

NIST® SRM® 2669

Sign Into View Organizational & Contract Pricing

Select a Size

10 VIALS
€1,180.00

€1,180.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 VIALS
€1,180.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€1,180.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

packaging

pkg of Set (10) (5 ea conc)

manufacturer/tradename

NIST®

technique(s)

ion chromatography: suitable
liquid chromatography (LC): suitable
mass spectrometry (MS): suitable

application(s)

clinical testing

format

matrix material

shipped in

dry ice

General description

SRM 2669 contains five pouches, with each consisting of one vial of Level I and another vial of Level II arsenic species in frozen human urine, each measuring approximately 1.5 mL of urine. For more information, please refer to the SDS and COA.
SRM 2669_cert


SRM 2669 _SDS

Application

This Standard Reference Material (SRM) is intended for the validation of analytical methods and measurements to determine arsenic species in human urine.

Features and Benefits

  • Accompanied by NIST-certified values for arsenic species that are metrologically traceable to the SI unit of mass concentration, expressed in micrograms per liter.
  • Non-certified values for total arsenic are provided.
  • Expiration details, along with instructions for handling, storage, and use, are included on the NIST certificate.

Preparation Note

SRM 2669 is shipped on dry ice, and it should be stored at –80 °C until use. The material should be used within 4 h after being thawed.

Other Notes

Certified for the analytes listed below.
Arsenous acid (H3AsO3), Arsenic acid (H3AsO4), Monomethylarsonic acid (CH5AsO3), Dimethylarsinic acid (C2H7AsO2), Trimethylarsine oxide (C3H9AsO), Arsenobetaine (C5H11AsO2), Arsenocholine (C5H14AsO)

Reference/Informational value is provided for the following analytes.
Arsenic (As, Trimethylarsine oxide (C3H9AsO), Arsenocholine (C5H14AsO)

See certificate for values and more details at nist.gov/SRM.
  • SRM 2669 is intended for research use
  • Details on expiration, storage, safety, usage, and source are provided in the NIST certificate
  • Information on biomaterials, disposal, and transport is available in the SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Clay Davis et al.
Analytical and bioanalytical chemistry, 396(8), 3041-3050 (2010-02-27)
Two independent liquid chromatography inductively coupled plasma-mass spectrometry (LC/ICP-MS) methods for the separation of arsenic species in urine have been developed with quantification by standard additions. Seven arsenic species have been quantified in a new NIST frozen human urine Standard
V M O Carioni et al.
Talanta, 165, 76-83 (2017-02-06)
The speciation analysis of As in urine samples can provide important information for epidemiological studies. Considering that these studies involve hundreds or thousands of samples, a fast and reliable method using a simple LC system with short-length mixed bed ion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service