Skip to Content
Merck
All Photos(1)

Documents

ERMDB001

Human hair (trace elements)

ERM®, certified reference material

Synonym(s):

Trace elements in human hair

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

format

neat

General description

Certified for As, Cd, Cu, Hg, Pb, Se, Zn.

Application

  • Assessment of trace element and mineral levels in students from Turkmenistan: This study compared the trace element and mineral profiles in hair samples from students in Turkmenistan with those from Iran and Russia, highlighting the utility of human hair as a biomarker in environmental exposure and nutritional status studies (Kirichuk et al., 2024).
  • Toxicological profile and potential health concerns in brick kiln workers: Research focused on metals and trace elements exposure in hair samples from brick kiln workers in Lahore, Pakistan, underlining human hair′s role in assessing occupational exposure and health risks (Saeed et al., 2024).
  • Iron, Copper, and Magnesium Concentration in Hair and Risk of Esophageal Cancer: A nested case-control study utilized hair samples to explore the relationship between trace elements and the risk of developing esophageal cancer, reinforcing the diagnostic potential of hair mineral analysis in clinical research (Hashemian et al., 2023).
  • Association of serum and hair antioxidant minerals with oxidative stress markers: This cross-sectional study examined the correlation between hair and serum antioxidant mineral levels and oxidative stress in healthy adults, demonstrating hair′s effectiveness as a biomarker for physiological and nutritional analysis (Kang et al., 2024).

Analysis Note

For more information please see:
ERMDB001

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriela P F Arrifano et al.
Frontiers in genetics, 9, 285-285 (2018-08-14)
Human exposure to mercury is a serious problem of public health in Amazon. As in other vulnerable populations throughout the world, Amazonian riverine populations are chronically exposed to this metal and some symptoms of mercury intoxication were already detected in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service