NIST2262
Chlorinated biphenyl cogeners in isooctane
NIST® SRM® 2262
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 5 x 1.2 mL
manufacturer/tradename
NIST®
application(s)
environmental
format
multi-component solution
General description
SRM 2262_cert
SRM 2262_SDS
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
71.6 °F
Flash Point(C)
22 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service