Skip to Content
Merck
All Photos(1)

Documents

NIST2259

PCB congeners in 2,2,4-trimethylpentane

NIST® SRM® 2259

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 ampules x 1.2 mL

manufacturer/tradename

NIST®

technique(s)

GC/MS: suitable

format

matrix material

General description

This Standard Reference Material (SRM) is a solution of 80 polychlorinated biphenyl (PCB) congeners in 2,2,4-trimethylpentane (isooctane). This SRM is intended primarily for use in the calibration of chromatographic instrumentation used for the determination of the certified compounds. A unit of SRM 2259 consists of five 2-mL ampoules, each containing approximately 1.2 mL of solution. For more information, please refer to the COA and SDS.

SRM 2259_cert

SRM 2259_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service