Skip to Content
Merck
All Photos(1)

Key Documents

NIST2257

PBDE congeners in 2,2,4-trimethylpentane

NIST® SRM® 2257

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 1.2 ML
€889.00

€889.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 X 1.2 ML
€889.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€889.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.2 mL

manufacturer/tradename

NIST®

format

matrix material

General description

Each unit of SRM 2257 contains five ampoules, each with a capacity of 2 mL, and holds ~ 1.2 mL of solution.


SRM 2257_cert

SRM 2257_SDS

Application

PBDE congeners in 2,2,4-trimethylpentane is intended to calibrate the chromatographic instrumentation used in PBDE congeners determination.

Features and Benefits

  • Accompanied with certificate documenting NIST-certified concentration values and estimated uncertainties for the select constituents in SRM2257.
  • The certified values are metrologically traceable to the International System of Units (SI) derived unit for mass fraction expressed as micrograms per gram.
  • The NIST certificate is provided with expiration certificate, storage, handling, use, and maintenance instructions.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service