Skip to Content
Merck
All Photos(1)

Key Documents

47927

Supelco

CEN PCB Congener Mix-1

10 μg/mL each component in heptane, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€71.10

€71.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€71.10

About This Item

UNSPSC Code:
12352200

€71.10


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

10 μg/mL each component in heptane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

CEN PCB congener mix-1 has been used as a standard in the identification and determination of polychlorinated biphenyl pesticide content, accumulated in samples of blood and viable eggs of sea turtles using gas chromatography method (GC). CEN PCB Congener Mix-1 is used as a certified reference standard for the identification and quantification of polychlorinated biphenyls (PCBs) in environmental matrices[1][2] using chromatography techniques.
CEN PCB congener mix-1 has been used as a standard in the identification and determination of polychlorinated biphenyl pesticide content, accumulated in samples of blood and viable eggs of sea turtles using gas chromatography method (GC).[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    2,2′,3,4,4′,5,5′-Heptachlorobiphenyl
    2,2′,3,4,4′,5′-Hexachlorobiphenyl
    2,2′,3,4′,5′,6-Hexachlorobiphenyl
    2,2′,4,4′,5,5′-Hexachlorobiphenyl
    2,2′,3,3′,4,4′,5,5′-Octachlorobiphenyl
    2,2′,4,5,5′-Pentachlorobiphenyl
    2,3′,4,4′,5-Pentachlorobiphenyl
    2,2′,3,5′-Tetrachlorobiphenyl
    2,2′,5,5′-Tetrachlorobiphenyl
    2,2′,5-Trichlorobiphenyl
    2,4,4′-Trichlorobiphenyl
    2,4′,5-Trichlorobiphenyl
    See All (12)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of organochlorine pesticides and PCB residues among hawksbill (Eretmochelys imbricata) and green (Chelonia mydas) turtles in the Yucatan Peninsula and their maternal transfer
Garcia-Besne G, et al.
Marine Pollution Bulletin, 91, 139-148 (2015)
Photochemical sample treatment for extracts clean up in PCB analysis from sediments
Barreca S, et al.
Talanta, 103, 349-354 (2013)
Simultaneous preconcentration of a wide variety of organic pollutants in water samples: Comparison of stir bar sorptive extraction and membrane-assisted solvent extraction
Prieto.A, et al.
Journal of Chromatography A, 1214(1-2), 1-10 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service