Skip to Content
Merck
All Photos(1)

Key Documents

C6888

Supelco

Cannabinol

analytical standard

Synonym(s):

CBN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O2
CAS Number:
Molecular Weight:
310.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

drug control

USDEA Schedule I; Home Office Schedule 1; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CCCCCc1cc(O)c-2c(OC(C)(C)c3ccc(C)cc-23)c1

InChI

1S/C21H26O2/c1-5-6-7-8-15-12-18(22)20-16-11-14(2)9-10-17(16)21(3,4)23-19(20)13-15/h9-13,22H,5-8H2,1-4H3

InChI key

VBGLYOIFKLUMQG-UHFFFAOYSA-N

Gene Information

human ... CNR2(1269)
rat ... Cnr1(25248)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Major metabolite of tetrahydrocannabinol that is not psychoactive, CB2 cannabinoid receptor agonist with immunosuppressive properties.

Other Notes

Component of marijuana from C. sativa

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J MacLennan et al.
British journal of pharmacology, 124(4), 619-622 (1998-08-05)
The cannabinoid receptor antagonist SR141716A has been suggested to be an inverse agonist at CB1 receptors in some isolated intact tissues. We found that the basal incorporation of [35S]-GTPgammaS in Chinese hamster ovary cells expressing human recombinant CB1 and CB2
A C Herring et al.
The Journal of pharmacology and experimental therapeutics, 291(3), 1156-1163 (1999-11-24)
Cannabinol (CBN), an immunosuppressive cannabinoid and ligand for the peripheral cannabinoid receptor CB2, inhibits the cAMP signaling cascade in forskolin-stimulated thymocytes. The objective of the present studies was to further characterize the mechanism of CBN immune modulation by investigating its
A C Herring et al.
Biochemical pharmacology, 55(7), 1013-1023 (1998-05-30)
Immune suppression by cannabinoids has been widely demonstrated in a variety of experimental models. The identification of two major types of G-protein-coupled cannabinoid receptors expressed on leukocytes, CB1 and CB2, has provided a putative mechanism of action for immune modulation
Lucio Minuti et al.
The Journal of organic chemistry, 77(18), 7923-7931 (2012-08-24)
Diels-Alder reactions of a range of 1-(alkoxy/alkyl/halogen-substituted phenyl)buta-1,3-dienes with methyl propiolate carried out in a green ethanolic medium under 9 kbar pressure were investigated. The use of high pressure as activating method of the Diels-Alder reactions allows efficient and regioselective
Jonathan A Farrimond et al.
Psychopharmacology, 223(1), 117-129 (2012-05-01)
Increased food consumption following ∆(9)-tetrahydrocannabinol-induced cannabinoid type 1 receptor agonism is well documented. However, possible non-∆(9)-tetrahydrocannabinol phytocannabinoid-induced feeding effects have yet to be fully investigated. Therefore, we have assessed the effects of the individual phytocannabinoids, cannabigerol, cannabidiol and cannabinol, upon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service