Skip to Content
Merck
All Photos(3)

Key Documents

C5070

Sigma-Aldrich

CHAPS hydrate

BioXtra, ≥98% (TLC)

Synonym(s):

3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, CHAPS

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€111.00
5 G
€378.00

€111.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€111.00
5 G
€378.00

About This Item

Empirical Formula (Hill Notation):
C32H58N2O7S · xH2O
CAS Number:
Molecular Weight:
614.88 (anhydrous basis)
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

€111.00


Please contact Customer Service for Availability

Request a Bulk Order

description

zwitterionic

Quality Level

product line

BioXtra

Assay

≥98% (TLC)

form

powder

mol wt

614.88 g/mol (anhydrous basis)

technique(s)

protein purification: suitable
protein quantification: suitable

impurities

≤0.001% Phosphorus (P)
≤0.1% Insoluble matter
<0.04% DMF

ign. residue

≤0.1%

mp

157 °C (dec.) (lit.)

solubility

H2O: 0.1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.C[C@H](CCC(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C32H58N2O7S.H2O/c1-21(8-11-29(38)33-14-6-15-34(4,5)16-7-17-42(39,40)41)24-9-10-25-30-26(20-28(37)32(24,25)3)31(2)13-12-23(35)18-22(31)19-27(30)36;/h21-28,30,35-37H,6-20H2,1-5H3,(H-,33,38,39,40,41);1H2/t21-,22+,23-,24-,25+,26+,27-,28+,30+,31+,32-;/m1./s1

InChI key

SJCUTFKCLFLIFE-JWTJKVBLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

CHAPS is a non-denaturing, zwitterionic sulfobetaine detergent. It is mild and solubilizes membrane proteins and receptors. It is useful for breaking protein-protein interactions. It is a derivative of the naturally occurring bile salts.[1] It has small micellar molecular weight (6,150) and high critical micellar concentration (6 - 10 mM) allowing it to be removed from samples by dialysis.

Application

  • 10mM CHAPS detergent has been used in the process of ion-exchange chromatography for improved purification.[2]
  • 0.5 w/v of CHAPS is a component of high salt buffer in protein purification alongside HEPES, NaCl and EDTA.[3]
  • HEPES buffer has been used as lysis buffer in immunoprecipitation, supplemented with 1% CHAPS.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yahya Homsi et al.
FEBS open bio, 7(2), 274-283 (2017-02-09)
Tetraspanins are cell membrane-scaffolding proteins interacting with one another and a repertoire of interaction partners. Through these interactions, they form extended molecular networks as tetraspanin webs or tetraspanin-enriched microdomains. Microscopic data suggest that these networks contain tetraspanin clusters, with poor
The specificity of homomeric clustering of CD81 is mediated by its d-loop.
Homsi Y and Lang T
FEBS Open Bio, 7(2), 274-283 (2017)
Arabidopsis thaliana IRX 10 and two related proteins from psyllium and P hyscomitrella patens are xylan xylosyltransferases.
Jensen JK, et al.
The Plant Journal, 80(2), 207-215 (2014)
Rongli Zhang et al.
Journal of molecular and cellular cardiology, 112, 64-73 (2017-09-09)
Heart failure is associated with mitochondrial dysfunction so that restoring or improving mitochondrial health is of therapeutic importance. Recently, reduction in NAD+ levels and NAD+-mediated deacetylase activity has been recognized as negative regulators of mitochondrial function. Using a cardiac specific
Jacqueline M Leung et al.
BMC molecular and cell biology, 21(1), 8-8 (2020-03-01)
TgDCX is a doublecortin-domain protein associated with the conoid fibers, a set of strongly curved non-tubular tubulin-polymers in Toxoplasma. TgDCX deletion impairs conoid structure and parasite invasion. TgDCX contains two tubulin-binding domains: a partial P25α and the DCX/doublecortin domain. Orthologues

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service